Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_017629.3 | NP_060099.2 | 16 | 2401 | |
XM_005270578.3 | XP_005270635.1 | 16 | 2286 | ||
XM_005270579.3 | XP_005270636.1 | 15 | 2094 | ||
XM_011540883.2 | XP_011539185.1 | 13 | 1890 | ||
XM_011540884.2 | XP_011539186.1 | 13 | 1753 | ||
GenBank mRNA | AB046787.1 | - | 16 | 2401 | |
AK000040.1 | - | 6 | 890 | ||
AL831908.1 | - | 15 | 2188 | ||
BC152450.1 | - | 16 | 2401 | 8862582 |