Gene Transcripts
- Gene Symbol
- ANTXRL
- Entrez Gene ID
- 195977
- Gene Name
- anthrax toxin receptor-like
- Gene Aliases
- Chromosome Location
- Chr. 10: 46286997 - 46330419 on Build GRCh38
- UniGene ID
- Hs.538515
- Species
- Homo sapiens
- Gene Symbol
- ANTXRLP1
- Entrez Gene ID
- 100996567
- Gene Name
- anthrax toxin receptor-like pseudogene 1
- Gene Aliases
- Chromosome Location
- Chr. 10: 46219025 - 46269608 on Build GRCh38
- UniGene ID
- Hs.546758
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_103827.1 |
|
2 |
273 |
|
GenBank mRNA |
BX280825.1 |
- |
2 |
241 |
|
HY092480.1 |
- |
2 |
273 |
|
View Full Product Details