Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001288629.1 | NP_001275558.1 | 7 | 1707 | |
NM_004440.3 | NP_004431.1 | 7 | 1707 | ||
XM_005248669.2 | XP_005248726.1 | 7 | 1764 | ||
XM_005248671.2 | XP_005248728.1 | 7 | 1764 | ||
XM_017010365.1 | XP_016865854.1 | 7 | 1764 | ||
GenBank mRNA | AB209269.1 | - | 7 | 1694 | |
AK313529.1 | - | 7 | 1679 | ||
BC126125.1 | - | 7 | 1581 | 8991841 | |
BC126151.1 | - | 7 | 1581 | 8991867 | |
BC143857.1 | - | 7 | 1581 | 9052373 | |
BC143858.1 | - | 7 | 1581 | 9052374 | |
L36642.1 | - | 7 | 1651 |