Gene Transcripts
- Gene Symbol
- ZBTB7C
- Entrez Gene ID
- 201501
- Gene Name
- zinc finger and BTB domain containing 7C
- Gene Aliases
- APM-1, APM1, ZBTB36, ZNF857C
- Chromosome Location
- Chr. 18: 48026886 - 48412639 on Build GRCh38
- UniGene ID
- Hs.515388
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001039360.2 |
NP_001034449.1
|
2, 3 |
1538 |
|
NM_001318841.1 |
NP_001305770.1
|
4, 5 |
1719 |
|
XM_005258229.4 |
XP_005258286.1
|
3, 4 |
1463 |
|
XM_011525861.2 |
XP_011524163.1
|
6, 7 |
1943 |
|
XM_011525863.2 |
XP_011524165.1
|
5, 6 |
2060 |
|
XM_011525864.2 |
XP_011524166.1
|
5, 6 |
1815 |
|
XM_011525865.2 |
XP_011524167.1
|
5, 6 |
2056 |
|
XM_011525866.2 |
XP_011524168.1
|
4, 5 |
1801 |
|
XM_011525867.2 |
XP_011524169.1
|
4, 5 |
1590 |
|
XM_011525869.2 |
XP_011524171.1
|
5, 6 |
2065 |
|
XM_011525870.2 |
XP_011524172.1
|
4, 5 |
1761 |
|
XM_011525871.2 |
XP_011524173.1
|
2, 3 |
2276 |
|
XM_017025605.1 |
XP_016881094.1
|
6, 7 |
3738 |
|
XM_017025606.1 |
XP_016881095.1
|
6, 7 |
2192 |
|
XM_017025607.1 |
XP_016881096.1
|
6, 7 |
1947 |
|
XM_017025608.1 |
XP_016881097.1
|
5, 6 |
3615 |
|
XM_017025609.1 |
XP_016881098.1
|
4, 5 |
1932 |
|
GenBank mRNA |
AM988836.1 |
- |
2, 3 |
1526 |
|
AM988837.1 |
- |
2, 3 |
1671 |
|
AM988838.1 |
- |
3, 4 |
1721 |
|
AM988839.1 |
- |
4, 5 |
1719 |
|
EF120357.1 |
- |
2, 3 |
1590 |
|
View Full Product Details