Gene Transcripts
- Gene Symbol
- SLC36A1
- Entrez Gene ID
- 206358
- Gene Name
- solute carrier family 36 member 1
- Gene Aliases
- Dct1, LYAAT1, PAT1, TRAMD3
- Chromosome Location
- Chr. 5: 151411344 - 151556318 on Build GRCh38
- UniGene ID
- Hs.269004
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001308150.1 |
NP_001295079.1
|
3, 4 |
458 |
|
NM_001308151.1 |
NP_001295080.1
|
3, 4 |
428 |
|
NM_078483.3 |
NP_510968.2
|
3, 4 |
458 |
|
XM_005268386.1 |
XP_005268443.1
|
3, 4 |
428 |
|
XM_011537580.2 |
XP_011535882.1
|
3, 4 |
1148 |
|
XM_011537581.1 |
XP_011535883.1
|
4, 5 |
576 |
|
XM_011537583.2 |
XP_011535885.1
|
4, 5 |
5013 |
|
XM_011537584.2 |
XP_011535886.1
|
3, 4 |
667 |
|
XM_011537585.1 |
XP_011535887.1
|
4, 5 |
523 |
|
XM_011537586.2 |
XP_011535888.1
|
3, 4 |
1271 |
|
XM_011537587.2 |
XP_011535889.1
|
3, 4 |
527 |
|
XM_011537588.2 |
XP_011535890.1
|
3, 4 |
371 |
|
XM_011537589.2 |
XP_011535891.1
|
3, 4 |
1279 |
|
XM_011537590.1 |
XP_011535892.1
|
3, 4 |
286 |
|
XM_011537591.1 |
XP_011535893.1
|
3, 4 |
261 |
|
XM_011537592.2 |
XP_011535894.1
|
3, 4 |
265 |
|
XM_011537595.2 |
XP_011535897.2
|
3, 4 |
1154 |
|
XM_011537596.2 |
XP_011535898.2
|
2, 3 |
240 |
|
XM_017009216.1 |
XP_016864705.1
|
3, 4 |
354 |
|
XM_017009217.1 |
XP_016864706.1
|
3, 4 |
458 |
|
XM_017009218.1 |
XP_016864707.1
|
2, 3 |
240 |
|
XM_017009220.1 |
XP_016864709.1
|
3, 4 |
428 |
|
GenBank mRNA |
AF516142.1 |
- |
2, 3 |
250 |
|
AK057340.1 |
- |
3, 4 |
428 |
|
AK309323.1 |
- |
3, 4 |
430 |
|
AY162213.1 |
- |
3, 4 |
363 |
|
AY227111.1 |
- |
4, 5 |
378 |
|
AY227113.1 |
- |
3, 4 |
334 |
|
BC136437.1 |
- |
3, 4 |
329 |
9020426 |
BC136438.1 |
- |
3, 4 |
329 |
9020427 |
BM787446.1 |
- |
3, 4 |
461 |
|
BX537963.1 |
- |
3, 4 |
455 |
|
GU296670.1 |
- |
2, 3 |
235 |
|
View Full Product Details