Gene Transcripts

Gene Symbol
SMTNL1
Entrez Gene ID
219537
Gene Name
smoothelin like 1
Gene Aliases
CHASM
Chromosome Location
Chr. 11: 57536841 - 57550274 on Build GRCh38
UniGene ID
Hs.68756
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105565.2 NP_001099035.2 1 234
XM_005273819.3 XP_005273876.1 2 1036

View Full Product Details