Gene Transcripts

Gene Symbol
C11orf45
Entrez Gene ID
219833
Gene Name
chromosome 11 open reading frame 45
Gene Aliases
Chromosome Location
Chr. 11: 128899565 - 128909520 on Build GRCh38
UniGene ID
Hs.689860
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256088.1 NP_001243017.1 4 1546
NM_145013.2 NP_659450.1 4 1535
NR_045767.1 4 1469
XM_011542654.2 XP_011540956.1 4 1465
XM_011542655.2 XP_011540957.1 4 1427
GenBank mRNA AK125634.1 - 4 1443
BC025756.1 - 4 1535 5201867

View Full Product Details