Gene Transcripts

Gene Symbol
OR5A2
Entrez Gene ID
219981
Gene Name
olfactory receptor family 5 subfamily A member 2
Gene Aliases
Chromosome Location
Chr. 11: 59421979 - 59422953 on Build GRCh38
UniGene ID
Hs.553648
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001954.1 NP_001001954.1 1 421
GenBank mRNA BC136991.1 - 1 444

View Full Product Details