Gene Transcripts

Gene Symbol
TSPO2
Entrez Gene ID
222642
Gene Name
translocator protein 2
Gene Aliases
BZRPL1
Chromosome Location
Chr. 6: 41041344 - 41044337 on Build GRCh38
UniGene ID
Hs.357392
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001010873.2 NP_001010873.1 4 704
NM_001159726.1 NP_001153198.1 4 514
XM_011514396.2 XP_011512698.1 4 507
XM_011514397.2 XP_011512699.1 4 1036
GenBank mRNA BC104440.2 - 3 489
BC104441.2 - 3 489
BC137560.1 - 4 484 9021560
BC137565.1 - 4 484 9021565
BC144710.1 - 4 514 9053248

View Full Product Details