Gene Transcripts

Gene Symbol
TCTEX1D2
Entrez Gene ID
255758
Gene Name
Tctex1 domain containing 2
Gene Aliases
Chromosome Location
Chr. 3: 196291219 - 196318294 on Build GRCh38
UniGene ID
Hs.733180
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152773.4 NP_689986.2 3 433
GenBank mRNA BC021177.2 - 3 413
BM548106.1 - 3 458
CK818899.1 - 3 427

View Full Product Details