Gene Transcripts

Gene Symbol
OR4C3
Entrez Gene ID
256144
Gene Name
olfactory receptor family 4 subfamily C member 3
Gene Aliases
OR11-98
Chromosome Location
Chr. 11: 48324941 - 48325930 on Build GRCh38
UniGene ID
Hs.553656
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004702.1 NP_001004702.1 1 322
GenBank mRNA BC136853.1 - 1 343
BC136854.1 - 1 343 9020845

View Full Product Details