Gene Transcripts

Gene Symbol
DSCR10
Entrez Gene ID
259234
Gene Name
Down syndrome critical region 10 (non-protein coding)
Gene Aliases
Chromosome Location
Chr. 21: 38206156 - 38208644 on Build GRCh38
UniGene ID
Hs.147695
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027695.1 3 577
GenBank mRNA AB066291.1 - 3 578
BC093855.1 - 1 227 7939700

View Full Product Details