Gene Transcripts

Gene Symbol
TAS2R39
Entrez Gene ID
259285
Gene Name
taste 2 receptor member 39
Gene Aliases
T2R39, T2R57
Chromosome Location
Chr. 7: 143183419 - 143184435 on Build GRCh38
UniGene ID
Hs.553660
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_176881.2 NP_795362.2 1 124
GenBank mRNA BC100947.2 - 1 124
BC100948.2 - 1 124
BC114952.1 - 1 124

View Full Product Details