Gene Transcripts

Gene Symbol
OR10AG1
Entrez Gene ID
282770
Gene Name
olfactory receptor family 10 subfamily AG member 1
Gene Aliases
OR11-160
Chromosome Location
Chr. 11: 55967558 - 55968463 on Build GRCh38
UniGene ID
Hs.554525
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005491.1 NP_001005491.1 1 589
GenBank mRNA BC136890.1 - 1 640
BC136891.1 - 1 640 9020882

View Full Product Details