Gene Transcripts
- Gene Symbol
- EYA1
- Entrez Gene ID
- 2138
- Gene Name
- EYA transcriptional coactivator and phosphatase 1
- Gene Aliases
- BOP, BOR, BOS1, OFC1
- Chromosome Location
- Chr. 8: 71197433 - 71548130 on Build GRCh38
- UniGene ID
- Hs.444971
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000503.5 |
NP_000494.2
|
5, 6 |
906 |
|
NM_172058.3 |
NP_742055.1
|
4, 5 |
498 |
|
NM_172059.3 |
NP_742056.1
|
4, 5 |
320 |
|
NM_172060.3 |
NP_742057.1
|
3, 4 |
370 |
|
XM_011517484.2 |
XP_011515786.2
|
6, 7 |
664 |
|
XM_017013201.1 |
XP_016868690.1
|
6, 7 |
692 |
|
XM_017013202.1 |
XP_016868691.1
|
6, 7 |
536 |
|
XM_017013205.1 |
XP_016868694.1
|
6, 7 |
664 |
|
XM_017013206.1 |
XP_016868695.1
|
7, 8 |
645 |
|
XM_017013207.1 |
XP_016868696.1
|
6, 7 |
663 |
|
XM_017013209.1 |
XP_016868698.1
|
6, 7 |
553 |
|
XM_017013211.1 |
XP_016868700.1
|
5, 6 |
536 |
|
XM_017013212.1 |
XP_016868701.1
|
4, 5 |
803 |
|
GenBank mRNA |
AF467247.1 |
- |
4, 5 |
295 |
|
AJ000097.1 |
- |
4, 5 |
443 |
|
AJ000098.1 |
- |
5, 6 |
904 |
|
BC121799.1 |
- |
4, 5 |
410 |
40084967 |
DC409057.1 |
- |
4, 5 |
553 |
|
View Full Product Details