Gene Transcripts

Gene Symbol
SLC39A5
Entrez Gene ID
283375
Gene Name
solute carrier family 39 member 5
Gene Aliases
LZT-Hs7, MYP24, ZIP5
Chromosome Location
Chr. 12: 56230036 - 56237846 on Build GRCh38
UniGene ID
Hs.591018
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135195.1 NP_001128667.1 7 1216
NM_173596.2 NP_775867.2 9 1245
XM_005268803.1 XP_005268860.1 7 1133
XM_011538198.1 XP_011536500.1 7 1198
XM_011538199.1 XP_011536501.1 9 1252
XM_011538200.1 XP_011536502.1 8 1202
XM_011538201.1 XP_011536503.1 8 1176
XM_017019185.1 XP_016874674.1 7 1176
XM_017019186.1 XP_016874675.1 9 1385
XM_017019187.1 XP_016874676.1 7 1134
GenBank mRNA AK172768.1 - 7 1179
AK313188.1 - 9 1208
BC027884.1 - 7 1090 5221329

View Full Product Details