Gene Transcripts

Gene Symbol
SMIM21
Entrez Gene ID
284274
Gene Name
small integral membrane protein 21
Gene Aliases
C18orf62
Chromosome Location
Chr. 18: 75409476 - 75455653 on Build GRCh38
UniGene ID
Hs.666744
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037331.3 NP_001032408.1 2, 3 396
GenBank mRNA AK093452.1 - 2, 3 396
BC107486.1 - 2, 3 327 40032395

View Full Product Details