Gene Transcripts

Gene Symbol
MAMSTR
Entrez Gene ID
284358
Gene Name
MEF2 activating motif and SAP domain containing transcriptional regulator
Gene Aliases
MASTR
Chromosome Location
Chr. 19: 48705718 - 48719898 on Build GRCh38
UniGene ID
Hs.191815
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130915.1 NP_001124387.1 10 1103
NM_001297753.1 NP_001284682.1 7 1216
NM_182574.2 NP_872380.1 8 1347
XM_011526807.2 XP_011525109.1 8 1352
XM_011526808.2 XP_011525110.1 10 1471
XM_011526809.2 XP_011525111.1 7 1150
XM_017026640.1 XP_016882129.1 9 1871
XM_017026641.1 XP_016882130.1 9 1832
XM_017026643.1 XP_016882132.1 9 1712
XM_017026644.1 XP_016882133.1 9 1682
XM_017026645.1 XP_016882134.1 6 1117
GenBank mRNA AK093389.1 - 8 1347
AK131427.1 - 10 1110
BC105056.1 - 6 771 8144059
BC105058.1 - 6 771 8144061
BC106047.1 - 7 1216 5786639
BC143443.1 - 5 576 9051951
BX280379.1 - 3 297

View Full Product Details