Gene Transcripts

Gene Symbol
FANCC
Entrez Gene ID
2176
Gene Name
Fanconi anemia complementation group C
Gene Aliases
FA3, FAC, FACC
Chromosome Location
Chr. 9: 95099054 - 95317730 on Build GRCh38
UniGene ID
Hs.494529
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000136.2 NP_000127.2 15 1849
NM_001243743.1 NP_001230672.1 15 1791
XM_005251802.3 XP_005251859.1 9 1951
XM_006717001.2 XP_006717064.1 14 1666
XM_011518365.2 XP_011516667.1 15 1824
XM_011518367.2 XP_011516669.1 16 1961
XM_017014452.1 XP_016869941.1 16 1976
XM_017014453.1 XP_016869942.1 16 1923
XM_017014454.1 XP_016869943.1 15 1850
GenBank mRNA AK312548.1 - 15 1714
BC015748.1 - 15 1839 4864916
CB052780.1 - 2 152
KU178051.1 - 14 1587
KU178052.1 - 15 1732
X66893.1 - 15 1839
X66894.1 - 15 1757

View Full Product Details