Gene Transcripts

Gene Symbol
C3orf56
Entrez Gene ID
285311
Gene Name
chromosome 3 open reading frame 56
Gene Aliases
Chromosome Location
Chr. 3: 127193131 - 127198185 on Build GRCh38
UniGene ID
Hs.591284
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007534.2 NP_001007535.1 2 1037
GenBank mRNA AK097460.1 - 2 1037
BC137154.1 - 2 998 9021151
BC137155.1 - 2 998 9021152

View Full Product Details