Gene Transcripts
- Gene Symbol
- NPEPPS
- Entrez Gene ID
- 9520
- Gene Name
- aminopeptidase puromycin sensitive
- Gene Aliases
- AAP-S, MP100, PSA
- Chromosome Location
- Chr. 17: 47522950 - 47623276 on Build GRCh38
- UniGene ID
- Hs.443837
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_006310.3 |
NP_006301.3
|
16, 17 |
2106 |
|
XM_011525496.1 |
XP_011523798.1
|
17, 18 |
1908 |
|
XM_017025372.1 |
XP_016880861.1
|
16, 17 |
2068 |
|
XM_017025373.1 |
XP_016880862.1
|
17, 18 |
1908 |
|
GenBank mRNA |
AJ132583.1 |
- |
16, 17 |
1836 |
|
AK096491.1 |
- |
1, 2 |
564 |
|
AK293450.1 |
- |
10, 11 |
1038 |
|
AK293995.1 |
- |
9, 10 |
1020 |
|
AK296887.1 |
- |
15, 16 |
1749 |
|
AK297107.1 |
- |
15, 16 |
1610 |
|
AK303037.1 |
- |
17, 18 |
1901 |
|
BC065294.1 |
- |
16, 17 |
1795 |
6059589 |
View Full Product Details