Gene Transcripts
- Gene Symbol
- OR2V2
- Entrez Gene ID
- 285659
- Gene Name
- olfactory receptor family 2 subfamily V member 2
- Gene Aliases
- OR2V3, OST713
- Chromosome Location
- Chr. 5: 181154943 - 181155890 on Build GRCh38
- UniGene ID
- Hs.553681
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_206880.1 |
NP_996763.1
|
1 |
219 |
|
- Gene Symbol
- OR2V1
- Entrez Gene ID
- 26693
- Gene Name
- olfactory receptor family 2 subfamily V member 1
- Gene Aliases
- OR2V1P, OST265
- Chromosome Location
- Chr. 5: 181124357 - 181125304 on Build GRCh38
- UniGene ID
- Hs.742585
- Species
- Homo sapiens
View Full Product Details