Gene Transcripts

Gene Symbol
KRTAP23-1
Entrez Gene ID
337963
Gene Name
keratin associated protein 23-1
Gene Aliases
KAP23.1
Chromosome Location
Chr. 21: 30348399 - 30348606 on Build GRCh38
UniGene ID
Hs.553688
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181624.1 NP_853655.1 1 56
GenBank mRNA BC128435.1 - 1 60

View Full Product Details