Gene Transcripts

Gene Symbol
OXLD1
Entrez Gene ID
339229
Gene Name
oxidoreductase like domain containing 1
Gene Aliases
C17orf90
Chromosome Location
Chr. 17: 81665036 - 81666637 on Build GRCh38
UniGene ID
Hs.405942
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039842.2 NP_001034931.1 2 604
NM_001304994.1 NP_001291923.1 2 958
NM_001304995.1 NP_001291924.1 2 845
NM_001304999.1 NP_001291928.1 1 1537
NR_130936.1 2 967
NR_130937.1 2 972
NR_130938.1 2 976
NR_130939.1 2 633
GenBank mRNA AY007126.1 - 1 1535
BC090923.1 - 2 555 30335041
CB048026.1 - 2 641

View Full Product Details