Gene Transcripts

Gene Symbol
PRR30
Entrez Gene ID
339779
Gene Name
proline rich 30
Gene Aliases
C2orf53
Chromosome Location
Chr. 2: 27136847 - 27139464 on Build GRCh38
UniGene ID
Hs.136555
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178553.3 NP_848648.2 3 804
GenBank mRNA BC045671.1 - 3 750

View Full Product Details