Gene Transcripts

Gene Symbol
SLC35D3
Entrez Gene ID
340146
Gene Name
solute carrier family 35 member D3
Gene Aliases
FRCL1
Chromosome Location
Chr. 6: 136922241 - 136925660 on Build GRCh38
UniGene ID
Hs.369703
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008783.2 NP_001008783.1 1 501
GenBank mRNA AK295424.1 - 1 350
AK316095.1 - 1 501
BC087842.1 - 1 478 6191889

View Full Product Details