Gene Transcripts

Gene Symbol
SATL1
Entrez Gene ID
340562
Gene Name
spermidine/spermine N1-acetyl transferase-like 1
Gene Aliases
Chromosome Location
Chr. X: 85092286 - 85116229 on Build GRCh38
UniGene ID
Hs.640783
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012980.2 NP_001012998.2 1 771
XM_011530940.2 XP_011529242.1 2 1205
GenBank mRNA BC043215.1 - 1 246
BC126401.1 - 1 231 8992117

View Full Product Details