Gene Transcripts

Gene Symbol
FCGRT
Entrez Gene ID
2217
Gene Name
Fc fragment of IgG receptor and transporter
Gene Aliases
FCRN, alpha-chain
Chromosome Location
Chr. 19: 49512279 - 49526428 on Build GRCh38
UniGene ID
Hs.111903
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001136019.2 NP_001129491.1 6 1443
NM_004107.4 NP_004098.1 5 1179
XM_005258657.1 XP_005258714.1 5 998
XM_011526627.2 XP_011524929.1 4 860
GenBank mRNA AK074734.1 - 2 2321
AK075510.1 - 6 1377
AK075532.1 - 6 1093
BC008734.2 - 5 1123 3163446
BE245284.1 - 2 213
BT006828.1 - 5 957
BX647163.1 - 6 1063
U12255.1 - 5 1082

View Full Product Details