Gene Transcripts

Gene Symbol
OR2W3
Entrez Gene ID
343171
Gene Name
olfactory receptor family 2 subfamily W member 3
Gene Aliases
OR2W3P, OR2W8P, OST718
Chromosome Location
Chr. 1: 247895587 - 247896531 on Build GRCh38
UniGene ID
Hs.269151
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001957.2 NP_001001957.2 1 766
GenBank mRNA BC146848.1 - 1 797

Gene Symbol
TRIM58
Entrez Gene ID
25893
Gene Name
tripartite motif containing 58
Gene Aliases
BIA2
Chromosome Location
Chr. 1: 247857199 - 247880138 on Build GRCh38
UniGene ID
Hs.269151
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BX537432.1 - 3 1023

View Full Product Details