Gene Transcripts
- Gene Symbol
- KIAA1211L
- Entrez Gene ID
- 343990
- Gene Name
- KIAA1211 like
- Gene Aliases
- C2orf55
- Chromosome Location
- Chr. 2: 98793846 - 98936221 on Build GRCh38
- UniGene ID
- Hs.469398
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_207362.2 |
NP_997245.2
|
4, 5 |
662 |
|
XM_005263923.3 |
XP_005263980.1
|
4, 5 |
642 |
|
XM_005263924.3 |
XP_005263981.1
|
4, 5 |
642 |
|
XM_005263926.3 |
XP_005263983.1
|
4, 5 |
527 |
|
XM_011511093.1 |
XP_011509395.1
|
6, 7 |
801 |
|
XM_011511094.1 |
XP_011509396.1
|
5, 6 |
699 |
|
XM_011511095.2 |
XP_011509397.1
|
4, 5 |
469 |
|
XM_017003971.1 |
XP_016859460.1
|
4, 5 |
645 |
|
XM_017003972.1 |
XP_016859461.1
|
5, 6 |
749 |
|
XM_017003973.1 |
XP_016859462.1
|
4, 5 |
655 |
|
GenBank mRNA |
BM922097.1 |
- |
4, 5 |
680 |
|
View Full Product Details