Gene Transcripts

Gene Symbol
RTP2
Entrez Gene ID
344892
Gene Name
receptor transporter protein 2
Gene Aliases
Z3CXXC2
Chromosome Location
Chr. 3: 187698259 - 187715625 on Build GRCh38
UniGene ID
Hs.567872
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004312.2 NP_001004312.2 2 1260
XM_017006301.1 XP_016861790.1 5 1454
XM_017006302.1 XP_016861791.1 5 1405
GenBank mRNA BC068081.1 - 2 1260

View Full Product Details