Gene Transcripts

Gene Symbol
BPIFB3
Entrez Gene ID
359710
Gene Name
BPI fold containing family B member 3
Gene Aliases
C20orf185, LPLUNC3, RYA3
Chromosome Location
Chr. 20: 33055331 - 33073637 on Build GRCh38
UniGene ID
Hs.472442
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_182658.2 NP_872599.1 14 1453
GenBank mRNA AF549189.1 - 14 1360
BC113398.1 - 14 1453 8322450
BC113400.1 - 14 1453 8322452

View Full Product Details