Gene Transcripts

Gene Symbol
VWC2
Entrez Gene ID
375567
Gene Name
von Willebrand factor C domain containing 2
Gene Aliases
PSST739, UNQ739
Chromosome Location
Chr. 7: 49773661 - 49923777 on Build GRCh38
UniGene ID
Hs.629302
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198570.4 NP_940972.2 4 1633
NR_136188.1 3 1180
GenBank mRNA AY358393.1 - 4 1657
BC110857.1 - 4 1576 5731863

View Full Product Details