Gene Transcripts

Gene Symbol
TMEM82
Entrez Gene ID
388595
Gene Name
transmembrane protein 82
Gene Aliases
Chromosome Location
Chr. 1: 15742492 - 15747797 on Build GRCh38
UniGene ID
Hs.454828
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013641.2 NP_001013663.1 6 1144
GenBank mRNA BC137239.1 - 6 1144
BC137240.1 - 6 1144 9021237

View Full Product Details