Gene Transcripts

Gene Symbol
TMPRSS11F
Entrez Gene ID
389208
Gene Name
transmembrane protease, serine 11F
Gene Aliases
Chromosome Location
Chr. 4: 68053198 - 68129869 on Build GRCh38
UniGene ID
Hs.677557
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207407.2 NP_997290.2 8 863
GenBank mRNA AK122625.1 - 8 863

View Full Product Details