Gene Transcripts

Gene Symbol
FLOT2
Entrez Gene ID
2319
Gene Name
flotillin 2
Gene Aliases
ECS-1, ECS1, ESA, ESA1, M17S1
Chromosome Location
Chr. 17: 28879339 - 28897728 on Build GRCh38
UniGene ID
Hs.514038
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004475.2 NP_004466.2 2 198
XM_005257950.3 XP_005258007.1 2 229
XM_005257953.2 XP_005258010.1 2 224
XM_017024394.1 XP_016879883.1 2 126
XM_017024395.1 XP_016879884.1 2 204
XM_017024396.1 XP_016879885.1 2 132
GenBank mRNA BC003683.1 - 2 110
BC017292.1 - 1 20
CA390100.1 - 2 198
M60922.1 - 2 54

View Full Product Details