Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001012276.3 | NP_001012276.2 | 4 | 1540 | |
XM_017001269.1 | XP_016856758.1 | 3 | 1536 | ||
GenBank mRNA | BC140755.1 | - | 4 | 1540 | |
BC144565.1 | - | 3 | 1542 | 9053093 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001012277.4 | NP_001012277.2 | 4 | 1540 |