Gene Transcripts

Gene Symbol
KAZN
Entrez Gene ID
23254
Gene Name
kazrin, periplakin interacting protein
Gene Aliases
KAZ
Chromosome Location
Chr. 1: 13893387 - 15118048 on Build GRCh38
UniGene ID
Hs.368823
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_201628.2 NP_963922.2 15 2491
XM_005245795.4 XP_005245852.1 17 2746
XM_011541074.2 XP_011539376.1 16 4682
XM_011541075.2 XP_011539377.1 14 4338
XM_011541076.2 XP_011539378.1 14 4483
XM_011541077.2 XP_011539379.1 14 5462
XM_011541081.2 XP_011539383.1 8 3162
GenBank mRNA AK293806.1 - 10 1297
BC101637.1 - 5 572
BC113621.1 - 5 572
BC143641.1 - 5 518
DA317442.1 - 5 509

View Full Product Details