Gene Transcripts
- Gene Symbol
- TRIM49D1
- Entrez Gene ID
- 399939
- Gene Name
- tripartite motif containing 49D1
- Gene Aliases
- TRIM49D, TRIM49D1P, TRIM49D2, TRIM49D2P, TRIM49DP
- Chromosome Location
- Chr. 11: 89911472 - 89920410 on Build GRCh38
- UniGene ID
- Hs.448674
- Species
- Homo sapiens
- Gene Symbol
- TRIM49D2
- Entrez Gene ID
- 729384
- Gene Name
- tripartite motif containing 49D2
- Gene Aliases
- TRIM48L1, TRIM49D1, TRIM49D2P, TRIM49L, TRIM49L1
- Chromosome Location
- Chr. 11: 89924064 - 89933063 on Build GRCh38
- UniGene ID
- Hs.705301
- Species
- Homo sapiens
View Full Product Details