Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001160227.1 | NP_001153699.1 | 15 | 2754 | |
NM_025137.3 | NP_079413.3 | 15 | 2754 | ||
XM_006720700.1 | XP_006720763.1 | 15 | 2754 | ||
XM_006720701.3 | XP_006720764.1 | 15 | 2754 | ||
XM_017022634.1 | XP_016878123.1 | 15 | 2754 | ||
XM_017022635.1 | XP_016878124.1 | 15 | 2754 | ||
XM_017022636.1 | XP_016878125.1 | 5 | 670 | ||
GenBank mRNA | AB058743.2 | - | 15 | 2724 | |
AB470308.1 | - | 15 | 2723 | ||
AB470309.1 | - | 15 | 2723 | ||
AB470310.1 | - | 15 | 2723 | ||
AL834168.1 | - | 14 | 2475 | ||
BC150640.1 | - | 15 | 2751 | 9057010 | |
BC153879.1 | - | 15 | 2724 | 8860108 |