Gene Transcripts
- Gene Symbol
- ATP2C1
- Entrez Gene ID
- 27032
- Gene Name
- ATPase secretory pathway Ca2+ transporting 1
- Gene Aliases
- ATP2C1A, BCPM, HHD, PMR1, SPCA1, hSPCA1
- Chromosome Location
- Chr. 3: 130850500 - 131016712 on Build GRCh38
- UniGene ID
- Hs.584884
- Species
- Homo sapiens
View Full Product Details