Gene Transcripts

Gene Symbol
TRIM37
Entrez Gene ID
4591
Gene Name
tripartite motif containing 37
Gene Aliases
MUL, POB1, TEF3
Chromosome Location
Chr. 17: 58968068 - 59106921 on Build GRCh38
UniGene ID
Hs.579079
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005207.3 NP_001005207.1 19 2517
NM_001320987.1 NP_001307916.1 18 2415
NM_001320988.1 NP_001307917.1 19 2517
NM_001320989.1 NP_001307918.1 19 2517
NM_001320990.1 NP_001307919.1 20 2593
NM_015294.4 NP_056109.1 19 2517
XM_005257385.1 XP_005257442.1 19 2349
XM_011524831.1 XP_011523133.1 19 2349
XM_011524832.2 XP_011523134.1 19 2349
XM_011524833.1 XP_011523135.1 18 2298
XM_011524834.1 XP_011523136.1 18 2231
XM_011524836.1 XP_011523138.1 19 2349
XM_017024662.1 XP_016880151.1 19 2349
XM_017024663.1 XP_016880152.1 19 2349
XM_017024664.1 XP_016880153.1 18 2298
XM_017024665.1 XP_016880154.1 18 2231
XM_017024666.1 XP_016880155.1 18 2298
XM_017024667.1 XP_016880156.1 18 2298
XM_017024668.1 XP_016880157.1 18 2231
XM_017024669.1 XP_016880158.1 17 2180
XM_017024670.1 XP_016880159.1 18 2230
XM_017024671.1 XP_016880160.1 18 2261
XM_017024672.1 XP_016880161.1 18 2261
XM_017024673.1 XP_016880162.1 18 2053
XM_017024674.1 XP_016880163.1 18 2053
GenBank mRNA AB020705.1 - 19 2102
AF213365.1 - 19 2321
AK022701.1 - 2 227
AK025648.1 - 13 1535
AK289674.1 - 19 2332
AK292459.1 - 19 2389
AK294850.1 - 18 2320
BC036012.1 - 19 2353 5270430
BX537955.1 - 20 2412

View Full Product Details