Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001024457.3 | NP_001019628.3 | 4 | 347 | |
XM_011532845.2 | XP_011531147.1 | 5 | 546 | ||
XM_011532849.2 | XP_011531151.1 | 5 | 546 | ||
XM_011532850.1 | XP_011531152.1 | 5 | 546 | ||
XM_011532851.2 | XP_011531153.1 | 5 | 546 | ||
XM_011532852.2 | XP_011531154.1 | 5 | 546 | ||
XM_017004103.1 | XP_016859592.1 | 5 | 546 | ||
XM_017004104.1 | XP_016859593.1 | 4 | 663 | ||
XM_017004105.1 | XP_016859594.1 | 4 | 511 | ||
XM_017004106.1 | XP_016859595.1 | 4 | 630 | ||
XM_017004107.1 | XP_016859596.1 | 5 | 546 | ||
GenBank mRNA | CR749330.1 | - | 4 | 347 | |
DC343949.1 | - | 4 | 422 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001078170.2 | NP_001071638.2 | 4 | 393 | |
XM_017004845.1 | XP_016860334.1 | 8 | 844 | ||
XM_017004846.1 | XP_016860335.1 | 4 | 515 | ||
XM_017004847.1 | XP_016860336.1 | 4 | 512 | ||
XM_017004848.1 | XP_016860337.1 | 8 | 844 | ||
XM_017004849.1 | XP_016860338.1 | 8 | 844 | ||
XM_017004850.1 | XP_016860339.1 | 8 | 844 | ||
GenBank mRNA | AK302429.1 | - | 4 | 393 |