Gene Transcripts

Gene Symbol
OR51A4
Entrez Gene ID
401666
Gene Name
olfactory receptor family 51 subfamily A member 4
Gene Aliases
Chromosome Location
Chr. 11: 4946159 - 4947100 on Build GRCh38
UniGene ID
Hs.555481
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005329.1 NP_001005329.1 1 447
GenBank mRNA BC140757.1 - 1 473

Gene Symbol
OR51A2
Entrez Gene ID
401667
Gene Name
olfactory receptor family 51 subfamily A member 2
Gene Aliases
Chromosome Location
Chr. 11: 4954772 - 4955713 on Build GRCh38
UniGene ID
Hs.553805
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004748.1 NP_001004748.1 1 447

View Full Product Details