Gene Transcripts

Gene Symbol
RPL23A
Entrez Gene ID
6147
Gene Name
ribosomal protein L23a
Gene Aliases
L23A, MDA20
Chromosome Location
Chr. 17: 28719982 - 28724356 on Build GRCh38
UniGene ID
Hs.419463
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000984.5 NP_000975.2 5 637
GenBank mRNA BC058041.1 - 5 633

View Full Product Details