Gene Transcripts

Gene Symbol
OR5K3
Entrez Gene ID
403277
Gene Name
olfactory receptor family 5 subfamily K member 3
Gene Aliases
Chromosome Location
Chr. 3: 98390666 - 98391631 on Build GRCh38
UniGene ID
Hs.554709
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005516.1 NP_001005516.1 1 574

View Full Product Details