Gene Transcripts

Gene Symbol
CUEDC1
Entrez Gene ID
404093
Gene Name
CUE domain containing 1
Gene Aliases
Chromosome Location
Chr. 17: 57861243 - 57955323 on Build GRCh38
UniGene ID
Hs.46679
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271875.1 NP_001258804.1 5, 6 1199
NM_001292025.1 NP_001278954.1 5, 6 1504
XM_006721900.2 XP_006721963.1 5, 6 1814
XM_011524812.2 XP_011523114.1 5, 6 587
XM_017024641.1 XP_016880130.1 6, 7 1216
GenBank mRNA AK000746.1 - 5, 6 1504
AK000977.1 - 4, 5 854
BC056882.1 - 5, 6 1199 6150503

View Full Product Details