Gene Transcripts

Gene Symbol
KRTAP5-5
Entrez Gene ID
439915
Gene Name
keratin associated protein 5-5
Gene Aliases
KRTAP5-11, KRTAP5.5
Chromosome Location
Chr. 11: 1629803 - 1630930 on Build GRCh38
UniGene ID
Hs.653467
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001480.2 NP_001001480.2 1 954
GenBank mRNA AJ628247.1 - 3 906

View Full Product Details