Gene Transcripts
- Gene Symbol
- PLEKHM1
- Entrez Gene ID
- 9842
- Gene Name
- pleckstrin homology and RUN domain containing M1
- Gene Aliases
- AP162, B2, OPTB6
- Chromosome Location
- Chr. 17: 45434262 - 45490780 on Build GRCh38
- UniGene ID
- Hs.514242
- Species
- Homo sapiens
- Gene Symbol
- PLEKHM1P1
- Entrez Gene ID
- 440456
- Gene Name
- pleckstrin homology and RUN domain containing M1 pseudogene 1
- Gene Aliases
- PLEKHM1P
- Chromosome Location
- Chr. 17: 64784841 - 64837184 on Build GRCh38
- UniGene ID
- Hs.646585
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024386.2 |
|
6 |
1469 |
|
GenBank mRNA |
AL833315.1 |
- |
6 |
1439 |
|
View Full Product Details