Gene Transcripts

Gene Symbol
RIMBP3
Entrez Gene ID
85376
Gene Name
RIMS binding protein 3
Gene Aliases
RIM-BP3, RIM-BP3.1, RIM-BP3.A, RIMBP3.1, RIMBP3A
Chromosome Location
Chr. 22: 18606124 - 18611919 on Build GRCh38
UniGene ID
Hs.115429
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015672.1 NP_056487.1 1 2633
GenBank mRNA AB051453.1 - 1 2005
AL133030.1 - 1 733

Gene Symbol
RIMBP3B
Entrez Gene ID
440804
Gene Name
RIMS binding protein 3B
Gene Aliases
RIM-BP3.2, RIM-BP3.B, RIMBP3.2
Chromosome Location
Chr. 22: 21383751 - 21389169 on Build GRCh38
UniGene ID
Hs.568257
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001128635.1 NP_001122107.1 1 2256

Gene Symbol
RIMBP3C
Entrez Gene ID
150221
Gene Name
RIMS binding protein 3C
Gene Aliases
RIM-BP3.3, RIMBP3.3
Chromosome Location
Chr. 22: 21545666 - 21551461 on Build GRCh38
UniGene ID
Hs.447643
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001128633.1 NP_001122105.1 1 2633

View Full Product Details